Learn More
Saint Gobain Life Sciences Tygon™ 2375 Ultra Chemical Resistant Tubing
Description
Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.
Tygon™ 2375 tubing from Saint-Gobain is ideal for applications where harsh cleaning chemicals are used. It is clear, flexible, and virtually unaffected by acids, bases, ketones, salts, and alcohols. This product is entirely free of plasticizers and eliminates fluid contamination, premature brittleness and cracking that often occurs with other flexible tubing products. When properly incinerated, Tygon™ 2375 does not release any pollution that could harm the environment.
- Outstanding chemical resistance
- Non-DEHP for high purity
- Plasticizer-free for low extractables
- Safer disposal – releases no harmful and corrosive hydrogen chloride gas
- Smoother inner surface – provides better flow and inhibits particulate buildup
- Low sorption – minimizes cross-contamination and maintains media purity
- Clear tubing for easier and better observation
Typical Applications
- Industrial/institutional cleaning
- Chemical transfer systems
- Transfer lines for chemical analyzers
- Detergent supply lines
- Ink and printing fluid dispensing
- Industrial X-ray processing
- Power generation sampling and drain lines
- Paint and solvent production
- Hazardous materials handling
Specifications
Specifications
| Diameter (English) Inner | 0.188 in. |
| Diameter (Metric) Inner | 4.8 mm |
| Material | Tygon |
| Diameter (English) Outer | 5/16 in. |
| Diameter (Metric) Outer | 7.9 mm |
| Overall Length (English) | 50 ft. |
| Overall Length (Metric) | 15 m |
| Thickness (English) | 0.0625 in. |
| Thickness (Metric) | 1.6 mm |
| Color | Clear |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.