missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ 2375 Ultra Chemical Resistant Tubing

Product Code. 11994438
Change view
Click to view available options
Diameter (English) Inner:
0.063 in.
0.188 in.
0.5 in.
0.625 in.
1/4 in.
5/16 in.
Diameter (English) Outer:
0.375 in.
0.75 in.
0.875 in.
3/16 in.
5/16 in.
7/16 in.
Thickness (Metric):
1.6 mm
3.2 mm
Thickness (English):
0.0625 in.
1/8 in.
Unit Size:
15m
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Diameter (English) Outer Thickness (Metric) Thickness (English) unitSize
11994438 1/4 in. 0.375 in. 1.6 mm 0.0625 in. 15m
11944438 0.063 in. 3/16 in. 1.6 mm 0.0625 in. 15m
11984438 0.188 in. 5/16 in. 1.6 mm 0.0625 in. 15m
11904448 5/16 in. 7/16 in. 1.6 mm 0.0625 in. 15m
11934448 0.5 in. 0.75 in. 3.2 mm 1/8 in. 15m
11944448 0.625 in. 0.875 in. 3.2 mm 1/8 in. 15m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11994438 Supplier Saint Gobain Life Sciences Supplier No. 9563016
  • ONLINE EXCLUSIVE PRICE
    216.00 €
    215.49 € / 15m
    Save 0.51 €
    0.24% Off
Estimated Shipment: 09-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tygon™ 2375 Ultra Chemical Resistant Tubing for Chemical Transfer Applications.

Use this tubing in highly regulated industries such as food and beverage, hospitality and institutional cleaning. Saint-Gobain™ Tygon™ Formula 2375 Chemical Resistant Tubing offers an unequaled combination of chemical resistance, clarity and flexibility.

Tygon™ 2375 tubing from Saint-Gobain is ideal for applications where harsh cleaning chemicals are used. It is clear, flexible, and virtually unaffected by acids, bases, ketones, salts, and alcohols. This product is entirely free of plasticizers and eliminates fluid contamination, premature brittleness and cracking that often occurs with other flexible tubing products. When properly incinerated, Tygon™ 2375 does not release any pollution that could harm the environment.

  • Outstanding chemical resistance
  • Non-DEHP for high purity
  • Plasticizer-free for low extractables
  • Safer disposal – releases no harmful and corrosive hydrogen chloride gas
  • Smoother inner surface – provides better flow and inhibits particulate buildup
  • Low sorption – minimizes cross-contamination and maintains media purity
  • Clear tubing for easier and better observation

Typical Applications

  • Industrial/institutional cleaning
  • Chemical transfer systems
  • Transfer lines for chemical analyzers
  • Detergent supply lines
  • Ink and printing fluid dispensing
  • Industrial X-ray processing
  • Power generation sampling and drain lines
  • Paint and solvent production
  • Hazardous materials handling

Specifications

Diameter (English) Inner 1/4 in.
Diameter (Metric) Inner 6.4 mm
Material Tygon
Diameter (English) Outer 0.375 in.
Diameter (Metric) Outer 9.5 mm
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.0625 in.
Thickness (Metric) 1.6 mm
Color Clear
Product Type Transfer Tubing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.