missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Sakuranetin

Product Code. 16469298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469298 Supplier MedChemExpress Supplier No. HYN3006
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Sakuranetin is a rice flavonoid phytoalexin, shows strong antifungal activity. Sakuranetin has anti-inflammatory and antioxidative activities. Sakuranetin ameliorates LPS-induced acute lung injury.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 286.28 g/mol
Shipping Condition Room temperature
For Use With (Application) COVID-19-immunoregulation
CAS 2957-21-3
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 125 mg/mL (436.64 mM)
Target Fungal
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.