missing translation for 'onlineSavingsMsg'
Learn More

Salicyl-AMS, MedChemExpress

Product Code. 16481119
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481119 1 mg 1mg
16476967 5 mg 5mg
16454248 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16481119 Supplier Medchem Express Supplier No. HY108941
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Salicyl-AMS is a mycobactin biosynthesis inhibitor which can also inhibit M. tuberculosis growth in vitro under iron-limited conditions.

Salicyl-AMS is a mycobactin biosynthesis inhibitor which can also inhibit M. tuberculosis growth in vitro under iron-limited conditions.

Specifications

Chemical Name or Material Salicyl-AMS
CAS 863238-55-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H18N6O8S
Quantity 1 mg
Solubility Information DMSO : ≥ 90.5 mg/mL (194.03 mM)
SMILES O[C@@H]1[C@@H](COS(=O)(NC(C2=C(O)C=CC=C2)=O)=O)O[C@@H](N3C(N=CN=C4N)=C4N=C3)[C@@H]1O
Molecular Weight (g/mol) 466.43
Formula Weight 466.43
Percent Purity 99.48%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.