missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, 99%

Product Code. 10367160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
100 g
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
1kg
250g
2g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367160 250 g Glass bottle 250g
10745791 2 g Glass bottle 2g
10071290 100 g Glass bottle 100g
10234900 1 kg Glass bottle 1kg
11944791 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10367160 Supplier Thermo Scientific Acros Supplier No. 132602500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 90-02-8 | C7H6O2 | 122.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-02-8 , 108-95-2
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00003317
InChI Key SMQUZDBALVYZAC-UHFFFAOYSA-N
Synonym salicylaldehyde, o-hydroxybenzaldehyde, o-formylphenol, salicylal, 2-formylphenol, benzaldehyde, 2-hydroxy, salicylic aldehyde, salicyladehyde, 2-hydroxy-benzaldehyde, benzaldehyde, o-hydroxy
PubChem CID 6998
ChEBI CHEBI:16008
IUPAC Name 2-hydroxybenzaldehyde
SMILES OC1=CC=CC=C1C=O

Specifications

Melting Point -7.0°C
Color Yellow
Density 1.1600g/mL
Boiling Point 197.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 1% max. (Phenol) (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CHO
Refractive Index 1.5720 to 1.5740
Quantity 250 g
Beilstein 08,31
Merck Index 15,8462
Specific Gravity 1.16
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, ether and benzene
Formula Weight 122.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Salicylaldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Salicylaldehyde
Signalwort
  • Danger||Warning
Gefahrenkategorie
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Gefahrenhinweis
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P391-Collect spillage.
  • P311-Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contain: Salicylaldehyde

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.