missing translation for 'onlineSavingsMsg'
Learn More

Salicylamide, TRC

Product Code. 30133936
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133936 10 g 10g
30133811 100 g 100g
30012158 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133936 Supplier Toronto Research Chemicals Supplier No. TRCS08809010G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-45-2
Molecular Formula C7 H7 N O2
Molecular Weight (g/mol) 137.14
Synonym Benzamide, 2-hydroxy-, Salicylamide (8CI), 2-Carbamoylphenol, 2-Carboxamidophenol, 2-Hydroxybenzamide, Acket, Afko-Sal, Algamon, Algiamida, Allevin, Amid-Sal, Amidosal, Anamid, Benesal, Cetamide, Cidal, Cymidon, Dropsprin, Liquiprin, Morsarinas, NSC 3115, NSC 83150, Novecyl, OHB, Oramid, Panithal, Raspberin, SR 4326, Salamid, Salamide, Saliamid, Saliamin, Salicim, Salicylic acid amide, Salipur, Salizell, Salrin, Salymid, Samid, Serramida, Urtosal, o-Hydroxybenzamide
IUPAC Name 2-hydroxybenzamide
SMILES NC(=O)c1ccccc1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H7NO2/c8-7(10)5-3-1-2-4-6(5)9/h1-4,9H,(H2,8,10)
Formula Weight 137.0477
Chemical Name or Material Salicylamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.