missing translation for 'onlineSavingsMsg'
Learn More

Salicylanilide, TRC

Product Code. 30428321
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428321 1 g -
30412483 250 mg 250mg
30412800 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428321 Supplier Toronto Research Chemicals Supplier No. TRCS0880951G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-17-2
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.23
Synonym 2-(N-Phenylcarboxamido)phenol, 2-(Phenylaminocarbonyl)phenol, 2-Hydroxy-N-phenylbenzamide, 2-Hydroxybenzanilide, Ansadol, Aseptolan, HyanilidN-Phenyl-2-hydroxybenzamide, N-Phenylsalicylamide, NSC 14881, SA 88, SA 88 (biocide), Salicylanilid, Salicylic acid anilide, Salifebrin, Salinide, Salinidol, Salnide, Sherstat SLN, Shirlan, Shirlan AG, Shirlan Extra, WR 10019, o-Hydroxybenzanilide
IUPAC Name 2-hydroxy-N-phenylbenzamide
SMILES Oc1ccccc1C(=O)Nc2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H11NO2/c15-12-9-5-4-8-11(12)13(16)14-10-6-2-1-3-7-10/h1-9,15H,(H,14,16)
Formula Weight 213.079
Chemical Name or Material Salicylanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.