missing translation for 'onlineSavingsMsg'
Learn More

Salicylic Acid, TRC

Product Code. 17942155
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17942155 250 mg 250mg
17901545 10 g 10g
17904685 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17942155 Supplier Toronto Research Chemicals Supplier No. TRCS088125250MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69-72-7
Molecular Formula C7 H6 O3
Molecular Weight (g/mol) 138.12
Synonym Salicylic acid, 2-Hydroxybenzoic acid, Lamivudine Imp. C (EP), Sulfasalazine Imp. H (EP), Carbasalate Calcium Imp. C (EP), Mesalazine Imp. H (EP), Acetylsalicylic Acid Imp. C (EP), Hydroxyethyl Salicylate Imp. A (EP), Benzoic acid, 2-hydroxy-, Salicylic acid (6CI,8CI), 2-Carboxyphenol, 2-Hydroxybenzenecarboxylic acid, Advanced Pain Relief Callus Removers, Advanced Pain Relief Corn Removers, Clear away Wart Remover, Compound W, Dr. Scholl's Callus Removers, Dr. Scholl's Corn Removers, Dr. Scholl's Wart Remover Kit, Duofil Wart Remover, Duoplant, Freezone, Ionil, Ionil Plus, K 537, K 557, NSC 180, Phenol-2-carboxylic acid, Psoriacid-S-Stift, Retarder W, Rutranex, Salicylic Acid Soap, Salicylic acid collodion, Saligel, Salonil, Salvona Nanosal, Stri-Dex, Trans-Ver-Sal, Verrugon, o-Carboxyphenol, o-Hydroxybenzoic acid
IUPAC Name 2-hydroxybenzoic acid
SMILES OC(=O)c1ccccc1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)
Formula Weight 138.0317
Percent Purity >95%
Chemical Name or Material Salicylic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.