missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Salmeterol-d3 (3-hydroxymethyl-d2; alpha-d1), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Salmeterol-d3 |
| CAS | 497063-94-2 |
| Molecular Formula | C25 2H3 H34 N O4 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C25H37NO4/c27-20-23-18-22(13-14-24(23)28)25(29)19-26-15-7-1-2-8-16-30-17-9-6-12-21-10-4-3-5-11-21/h3-5,10-11,13-14,18,25-29 H,1-2,6-9,12,15-17,19-20H2/i20D2,25 D |
| SMILES | [2 H]C([2 H])(O)c1cc(ccc1O)C([2 H])(O)CNCCCCCCOCCCCc2ccccc2 |
| IUPAC Name | 4-[1-deuterio-1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]-2-[dideuterio(hydroxy)methyl]phenol |
| Molecular Weight (g/mol) | 418.58 |
| Formula Weight | 418.2911 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction