missing translation for 'onlineSavingsMsg'
Learn More

Salsalate, TRC

Product Code. 17973315
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17973315 100 mg 100mg
17960925 50 mg 50mg
17924425 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973315 Supplier Toronto Research Chemicals Supplier No. TRCS095700100MG
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-94-3
Molecular Formula C14 H10 O5
Molecular Weight (g/mol) 258.23
Synonym 2-[(2-Hydroxybenzoyl)oxy]benzoic Acid, Carbasalate Calcium Imp. D (EP), Acetylsalicylic Acid Imp. E (EP), Salicylsalicylic Acid
IUPAC Name 2-(2-hydroxybenzoyl)oxybenzoic acid
SMILES OC(=O)c1ccccc1OC(=O)c2ccccc2O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H10O5/c15-11-7-3-1-5-9(11)14(18)19-12-8-4-2-6-10(12)13(16)17/h1-8,15H,(H,16,17)
Formula Weight 258.0528
Percent Purity >95%
Chemical Name or Material 2-[(2-Hydroxybenzoyl)oxy]benzoic Acid (Salicylsalicylic Acid)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.