missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Salubrinal

Product Code. 18014502
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18014502 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18014502 Supplier R&D Systems™ Supplier No. 2347/10
  • ONLINE EXCLUSIVE PRICE
    321.26 €
    287.70 € / 10mg
    Save 33.56 €
    10% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of eIF2α dephosphorylation

Cell-permeable, selective inhibitor of cellular phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF2α). Protects cells from endoplasmic reticulum stress-induced apoptosis (EC50 ∽ 15 μM).
TRUSTED_SUSTAINABILITY

Specifications

CAS 405060-95-9
Quantity 10 mg
Product Line Tocris
Molecular Formula C21H17Cl3N4OS
Formula Weight 479.81
Purity >99%
Molecular Weight (g/mol) 479.804
Chemical Name or Material Salubrinal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.