missing translation for 'onlineSavingsMsg'
Learn More

Salvequick Pilferproof Plaster Refill

Product Code. 12920475
Change view
Click to view available options
Material:
Fabric
Metal-detectable
Plastic
Color:
Beige
Blue
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Color Unit Size
12920475 Plastic Beige -
12940475 Metal-detectable Blue Each
12930475 Fabric Beige Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12920475 Supplier Safety First Supplier No. CD5
  • ONLINE EXCLUSIVE PRICE
    34.85 €
    28.93 € / Pack of 1
    Save 5.92 €
    16% Off
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Material Plastic
Product Type Salvequick Plaster Refill
Sterility Sterile
Quantity 6 Pk.
Dimensions (L x W) 7.2 x 1.9 cm/7.2 x 2.5 cm
Packaging Type Refill
For Use With (Application) Salvequick Pilferproof Plaster Dispenser, Food Industry
Color Beige
Includes Twenty-seven 72 x 19 mm; Eighteen 72 x 25 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.