missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) bromide hexahydrate, 99.99%, Thermo Scientific™

Product Code. 11443717
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443717 10 g 10g
11453717 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11443717 Supplier Thermo Scientific Alfa Aesar Supplier No. A18191.09
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13517-12-9
Molecular Formula Br3H12O6Sm
Molecular Weight (g/mol) 498.162
MDL Number MFCD00192430
InChI Key LXJDCZVUWJNXRL-UHFFFAOYSA-K
Synonym acmc-20ajrk, samarium bromide hexahydrate, samarium 3+ bromide-water 1/3/6, samarium iii bromide hexahydrate, reacton™
PubChem CID 57348178
IUPAC Name samarium(3+);tribromide;hexahydrate
SMILES O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]

Specifications

Melting Point >640°C
Quantity 10 g
Formula Weight 498.15 (390.06 Anhydrous)
Percent Purity 99.99%
Sensitivity Hygroscopic
Chemical Name or Material Samarium(III) bromide hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.