missing translation for 'onlineSavingsMsg'
Learn More

Samarium ingot, 99.9% (REO)

Product Code. 11396507
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396507 100 g 100g
11386507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396507 Supplier Thermo Scientific Alfa Aesar Supplier No. 000295.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7440-19-9 | Sm | 150.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 7440-19-9
Molecular Formula Sm
Molecular Weight (g/mol) 150.36
MDL Number MFCD00011233 MFCD00151299
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
PubChem CID 23951
ChEBI CHEBI:33374
IUPAC Name samarium
SMILES [Sm]

Spezifikation

Color Silver to Gray
Appearance Silver Gray solid
Quantity 100 g
Odor Odorless
Assay Percent Range 99.9% (REO)
Chemical Name or Material Samarium, ingot

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.