missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One Sample Cup for Coulter Counter™ Analyzers

Product Code. 10550752
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 1250
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10550752 Clear Pack of 1250
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10550752 Supplier Greiner Bio-One Supplier No. 668102
  • ONLINE EXCLUSIVE PRICE
    189.00 €
    146.54 € / Pack of 1250
    Save 42.46 €
    22% Off
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Coulter Counter Analyzer systems. Greiner Bio-One Sample Cup for Coulter Counter™ Analyzers are made of polystyrene.

  • Supplied with lid
  • Flat bottom
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Coulter Counter analyzer
Color Clear
Material Polystyrene
Product Type Analyzer Cup
Volume (Metric) 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.