missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Sample Grinding Kit

Product Code. 10084634 Shop All Cytiva Products
Click to view available options
Quantity:
50 Samples
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10084634

Brand: Cytiva 80648337

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Microcentrifuge tubes with grinding resin, 50; Pestles, 50

Disrupts small tissue and cell samples for protein extraction

  • Uses 1.5mL microcentrifuge tubes, grinding resin, and disposable pestles
  • Process up to 100mg of sample per tube in 10 minutes

TRUSTED_SUSTAINABILITY

Specifications

Product Type Sample grinding kit
Quantity 50 Samples
Includes 1.5mL Microcentrifuge tubes prepacked with grinding Resin, disposable pestles
Content And Storage Room Temperature
For Use With (Application) Mechanical breaking up of cells or tissue releasing the to tal Protein content
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.