missing translation for 'onlineSavingsMsg'
Learn More

Labcold™ Sample and Reagent Refrigerator

Product Code. 15863651
Change view
Click to view available options
Capacity (Metric):
150 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15863651 150 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15863651 Supplier Labcold™ Supplier No. RLDF0510A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Superior temperature uniformity with improved insulation for greater energy efficiency.

  • Fitted with an IntelliCold™ controller
  • Self-closing door
  • Door lock
  • Digital temperature control
  • Digital temperature display
  • Internal light
  • High/low temperature alarm
  • Min/max temperature recording
  • Automatic temperature and fridge function data recording
  • Door open alarm
  • Mains failure alarm
  • Alarm battery back-up
  • Off cycle auto defrost
  • Fan-assisted cooling
  • Remote alarm contacts
  • Access port
  • Low GWP refrigerant
  • Ammonia free
  • DH Green Book Guidelines compliant
  • Five-year warranty

Specifications

Capacity (Metric) 150 L
Temperature Range 2°C to 8°C
Door Style Solid
Hinge Side Right
Shelves 3
Exterior Finish Powder-coated Steel
Dimensions (DxWxH) Exterior 835 x 600 x 600 mm
Dimensions (D x W x H) Interior 690 x 480 x 410 mm
Weight (Metric) 51 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.