Learn More
Thermo Scientific™ Sample Sensor for Thermo Scientific™ Heratherm™ Advanced Protocol Security Incubators
Product Code. 10600411 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Sensor
Unit Size:
Each
Product Code. Type Unit Size
10600411 Sensor Each
1 options
Product Code. 10600411 Supplier Thermo Scientific™ Supplier No. 50127768
  • ONLINE EXCLUSIVE PRICE
    340.37 €
    306.00 € / Pack of 1
    Save 34.37 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Measures exact temperature
  • For connection to all Advanced Protocol Security incubators
  • Measures exact sample temperature
  • Temperature shown on display as plugged in
  • Cable length: 86.6 in.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Advanced Protocol Security Incubators
Type Sensor