Learn More
Buerkle™ Stainless Steel Sample Spoon
Buerkle™ Stainless Steel (AISI 304) Sample Spoon is a specially shaped spoon that allows you to reach almost every corner in a container and prevents samples from trickling out.
8.40 € - 24.85 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10297412
|
Buerkle™
5320-0210 |
210 mm |
10.10 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10494612
|
Buerkle™
5320-0300 |
300 mm |
14.05 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10552733
|
Buerkle™
5320-0150 |
150 mm |
8.95 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
10562733
|
Buerkle™
5320-0180 |
180 mm |
8.40 €
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
10044162
|
Buerkle™
5320-0400 |
400 mm |
21.45 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16369541
|
Buerkle™
5320-0500 |
500 mm |
24.85 €
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
- Made of AISI 304 (1.4301) stainless steel
- Largely chemical resistant (not against acids and alkalis)
Specifications
Stainless Steel (V2A) | |
Autoclavable |