missing translation for 'onlineSavingsMsg'
Learn More

Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh, Fisher Chemical™

Product Code. 10336100 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
3 kg
Unit Size:
1kg
3kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10336100 1 kg 1kg
10519770 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336100 Supplier Fisher Chemical Supplier No. S/0360/60
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sand, Extra Pure, SLR, Low Iron, 40-100 Mesh

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230°C
Melting Point 1610°C
Color Beige
Physical Form Solid
Quantity 1 kg
Formula Weight 60.08g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.