missing translation for 'onlineSavingsMsg'
Learn More

Sand, pure, 40-100 mesh

Product Code. 10602433 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Plastic bottle
Unit Size:
1kg
250g
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10602433 5 kg Plastic bottle 5kg
10267003 250 g Plastic bottle 250g
10692423 1 kg Plastic bottle 1kg
15224714 25 kg Fiber Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10602433 Supplier Thermo Scientific Acros Supplier No. 370940050
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14808-60-7 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
SMILES O=[Si]=O

Specifications

Boiling Point 2230.0°C
Melting Point 1610.0°C
Color Beige to Gray
Physical Form Crystals
Quantity 5 kg
Packaging Plastic bottle
Linear Formula SiO2
Merck Index 13,8567
Solubility Information Solubility in water: insoluble.
IUPAC Name dioxosilane
Formula Weight 60.08
Grade Pure
Chemical Name or Material Sand, 40 to 100 Mesh
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.