missing translation for 'onlineSavingsMsg'
Learn More

Sand, White Quartz, Honeywell Fluka™

Catalog Number 15697260
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Poly bottle
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O
14808-60-7
O2Si
60.08
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
24261
CHEBI:30563
dioxosilane
O=[Si]=O

Purum p.a., powder

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Boiling Point 2210°C
Melting Point 1710°C
Color White
Physical Form Solid
Quantity 1 kg
Linear Formula SiO2
UN Number NONH for all modes of transport
Formula Weight 60.08g/mol
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Sand, white quartz
Show More Show Less
Product Identifier
  • Sand, white quartz
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'