missing translation for 'onlineSavingsMsg'
Learn More

Sanguinarine chloride, Thermo Scientific™

Product Code. 17750639 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17750639 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17750639 Supplier Thermo Scientific Acros Supplier No. 468962500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Color Light Yellow to Dark Orange
Description Sanguinarine chloride
CAS 5578-73-4
Molecular Formula C20 H14 Cl N O4
Shipping Condition No special requirements
Product Line Thermo Scientific
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.