Learn More
Cole-Parmer Sanitary Clamps, GL Nylon
Product Code. 30052965 Shop All Cole-Parmer Products
Change view
Click to view available options
Includes:
1-1/2 in. Sanitary Clamp Size
2 in. Sanitary Clamp Size
3 in. Sanitary Clamp Size
3/4 in. Sanitary Clamp Size
4 in. Sanitary Clamp Size
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
30052965 1-1/2 in. Sanitary Clamp Size Each
30053842 3 in. Sanitary Clamp Size Each
30053650 4 in. Sanitary Clamp Size Each
30053689 3/4 in. Sanitary Clamp Size Each
30054086 2 in. Sanitary Clamp Size Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30052965 Supplier Cole-Parmer Supplier No. 5011882
  • ONLINE EXCLUSIVE PRICE
    29.58 €
    24.97 € / Pack of 1
    Save 4.61 €
    15% Off
Estimated Shipment: 05-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-quality nonmetallic sanitary clamps for critical fluid applications
Meet US and EU standards for use in pharmaceutical process applications. ASME-BPE, FDA, and USP Class VI compliant. Lightweight and reusable. Rust and corrosion resistant. Compatible with Tri-Clamp™ fittings. Sterilize by autoclave or gamma irradiation. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance ADCF;FDA;RoHS;USP
Min. Operating Temperature (Metric) -40°F
Packaging Quantity 100/Pk.
Sterilization Method Autoclave, gamma irradiation
Includes 1-1/2 in. Sanitary Clamp Size