Learn More
Cole-Parmer Sanitary Pinch Clamp, Nylon
Product Code. 30053749 Shop All Cole-Parmer Products
Click to view available options
Material:
Nylon
Unit Size:
Each
Product Code. 30053749
Brand: Cole-Parmer 5011914
This item requires a price update prior to being ordered.
Request a Price for this item here.
Designed to handle higher pressures than standard pinch clamps
Lightweight and reusable. Rust and corrosion resistant. USP Class VI compliant—excellent for biotechnology and pharmaceutical applications for transferring sterile fluids. Sterilize by autoclave or gamma irradiation. Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system. Free technical application support available to determine which connector type is needed for your application
Specifications
Certifications/Compliance FDA;USP
Material Nylon
Min. Operating Temperature (Metric) 0°C
Sterilization Method Autoclave, gamma irradiation
Color White