Learn More
Skytec™ Sapphire Zinc Gloves
Product Code. 30274502
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Unit Size:
10 pairs
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30274502 Small 10 pairs
30274521 Medium 10 pairs
30274621 Large 10 pairs
30274612 X-Large 10 pairs
30274623 2X-Large 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 30274502 Supplier Skytec™ Supplier No. SKG00086FD
  • ONLINE EXCLUSIVE PRICE
    70.81 €
    46.82 € / 10 pairs
    Save 23.99 €
    33% Off
In Stock
Add to basket
Edge
Add to basket
Glass and steel fiber free uncoated gloves offering high cut protection. Can be used as an under glove or worn by itself.
  • High Cut Resistant Uncoated Glove Made with Engineered HPPE
  • Cut resistant: Level E / A9
  • Engineered HPPE offers outstanding cut protection
  • Glass and steel fiber free reduce risks of irritation
  • Spandex in the liner allows the glove to fit the hand perfectly
  • Designed for easy movement and continuous wear
  • Breathable to reduce perspiration
  • Seamless knit prevents irritation
  • Elasticated wrist provides a secure fit
  • Ambidextrous for easy and quick donning
Specifications
Certifications/Compliance (Europe) CE II, UKCA, ANSI/ISEA 105-2016: Cut A5, EN 388:2016 + A1:2018 2X43E, EN 21420:2020
Coating Material Uncoated
Color Gray
Cut Resistance Rating Level E
Hand Ambidextrous
Latex Free Yes
Material HPPE
Product Type Cut-resistant Gloves
Size Small
EN388 Standard 2X43E
Length (Metric) 230 mm