missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

Product Code. 10235630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10235630 100 g Plastic bottle 100g
10041860 500 g Plastic bottle 500g
11905341 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10235630 Supplier Thermo Scientific Chemicals Supplier No. 157741000
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym N-Methylglycine
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Color Colorless or White to Yellow
Physical Form Crystals or Powder
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic bottle
Assay Percent Range 98%
Linear Formula CH3NHCH2CO2H
Water 2% max.
Formula Weight 89.09
Percent Purity ≥97.5% (total base)
Chemical Name or Material Sarcosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.