missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine, 98%

Product Code. 11432018
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432018 500 g 500g
11422018 100 g 100g
11442018 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432018 Supplier Thermo Scientific Chemicals Supplier No. A14594.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-97-1 | C3H7NO2 | 89.09 g/mol

A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A type 1 glycine transporter inhibitorSarcosine is used in the production of biodegradable surfactants and toothpastes. It is used as an intermediate in the synthesis of antienzyme agents. It is found to be a marker for prostate cancer bioagression. It acts as a biochemical reagent, dye stabilizer, daily chemical, amino acid surfactants, health drugs fatigue recovery agent and so on. It is involved in the preparation of N-methyl-glycine ethyl ester by reacting with ethanol.

Solubility
Soluble in water, methanol. Slightly soluble in ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00004279
InChI Key FSYKKLYZXJSNPZ-UHFFFAOYSA-N
Synonym sarcosine, n-methylglycine, sarcosinic acid, methylglycine, glycine, n-methyl, methylaminoacetic acid, n-methylaminoacetic acid, sarcosin, 2-methylamino acetic acid, methylamino acetic acid
PubChem CID 1088
ChEBI CHEBI:15611
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(O)=O

Specifications

Melting Point ∼208°C (decomposition)
pH 6.1
Odor Odorless
Quantity 500 g
Beilstein 1699442
Sensitivity Hygroscopic
Merck Index 14,8373
Solubility Information Soluble in water,methanol. Slightly soluble in ether.
Formula Weight 89.09
Percent Purity 98%
Chemical Name or Material Sarcosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.