missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine, TRC

Product Code. 30011147
Change view
Click to view available options
Quantity:
50 mg
25 g
100 g
Unit Size:
100g
25g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011147 25 g 25g
30004363 50 mg 50mg
30008126 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011147 Supplier Toronto Research Chemicals Supplier No. TRCS14050025G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-97-1
Molecular Formula C3 H7 N O2
Molecular Weight (g/mol) 89.09
Synonym N-Methylglycine, Sarcosine (8CI), N-Methylglycine, (Methylamino)acetic acid, (Methylamino)ethanoic acid, 2-(Methylamino)acetic acid, Acetic acid, (methylamino)-, Methylglycine, N-Methylaminoacetic acid, Sarcosin, Sarcosinic acid
IUPAC Name 2-(methylamino)acetic acid
SMILES CNCC(=O)O

Specifications

InChI Formula InChI=1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)
Quantity 25 g
Formula Weight 89.0477
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material Sarcosine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.