missing translation for 'onlineSavingsMsg'
Learn More

Sarkosyl, Fisher BioReagents

Product Code. 10698963
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10698963

Brand: Fisher BioReagents BP234500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Sarkosyl is an anionic detergent used for cell lysis during the extraction of RNA
  • Also suitable for the solubilization of membrane protein
TRUSTED_SUSTAINABILITY

Specifications

Color White
CAS 137-16-6
Physical Form Solid
Melting Point 45°C
pH 7.5 to 8.5
Quantity 500 g
Identification Pass Test
Packaging Poly Bottle
Assay Percent Range ≥95 %
Molecular Formula C15H28NNaO3
Linear Formula NaO2CCH2N(CH3)CO(CH2)10CH3
MDL Number MFCD00042728
Merck Index 15, 4401
Synonym Sodium Lauroyl Sarcosinate
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
Molecular Weight (g/mol) 293.38
PubChem CID 23668817
Formula Weight 293.38
Percent Purity ≥95%
Chemical Name or Material Sarkosyl
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Lauroylsarcosine sodium salt
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.