missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (BA.2.9.1; BA.2.13) His-tag Recombinant Protein

Product Code. 17936281
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 17936281

Brand: Invitrogen™ RP88552

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. The mutations were identified in the SARS-CoV-2 varient known as BA.2.9.1 or BA.2.13

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control
Format Lyophilized
Formulation PBS with 0.01% mannitol, 0.01% Tween 80, 5-8% trehalose and no preservative; pH 7.4
Molecular Weight (g/mol) 35.43 kDa
Name SARS-CoV-2 Spike Protein (RBD) (BA0.2.9.1; BA0.2.13) His-tag
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein RBD
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SARS-CoV-2 spike RBD (Accession # YP_009724390.1), amino acids Arg319-Phe541 with G339D, S371F, S373P, S375F, T376A, D405N, R408S, K417N, N440K, L452M, S477N, T478K, E484A, Q493R, Q498R, N501Y, Y505H substitutions and a C-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >90% as determined by SDS-PAGE. >90% as determined by SEC-HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.