missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ SARS-CoV Antigen Microarray

Product Code. 16317768
Change view
Click to view available options
Quantity: 1 Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16317768 Quantity: 1 Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16317768 Supplier PEPperCHIP™ Supplier No. SPC.019.001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SARS-CoV Antigen Microarray with 998 antigen specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

  • Microarray Content: 4 selected antigens of SARS coronavirus: spike glycoprotein (S protein), nucleoprotein (Nucleocapsid protein, NC), M protein, envelope small membrane protein (E protein, sM)
  • Peptide Length/Overlap: 15 aa/ 13 aa
  • Number of Peptides: 998 printed in duplicate and HA, polio and c-Myc control peptides

Spezifikation

Coating PEGMA/PMMA
Description 3 array copies per microarray
Quantity 1 Slide
Method of Attachment Combinatorial peptide synthesis by amino acid laser printing
Thickness (Metric) 1 mm
Certifications/Compliance ISO 9001:2015
Color Transparent
Regulatory Status RUO
Barcode Individual barcode for each microarray
Material Microscopic Glass
Type High-density Peptide Microarray
For Use With (Application) Antibody validation, epitope mapping, serum screening, biomarker discovery
Sterility Non-sterile
Dimensions (LxW) 75.4 x 25 mm
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.