Learn More
Sartorius Sartobind™ Lab LC System Adapter Kit
GREENER_CHOICE
Product Code. 30178673 Shop All Sartorius Products
Change view
Click to view available options
Material:
PEEK (Frit)
Unit Size:
1 pack
Product Code. Material Unit Size
30178673 PEEK (Frit) 1 pack
1 options
Product Code. 30178673 Supplier Sartorius Supplier No. SBLAAU011
  • ONLINE EXCLUSIVE PRICE
    22.40 €
    21.30 € / Pack of 1
    Save 1.10 €
    4% Off
In Stock
Add to basket
Add to basket
Easily convert the Luer connectors on any Sartobind™ Lab unit to the UNF 10-32 standard.
  • High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes.
  • Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity.
  • Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration.
  • Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system.
  • Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production.
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Material PEEK (Frit)