missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Sartobind™ Rapid A Lab
BIOPHARMA

Product Code. 30178672 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
1 Unit
4 Units
Unit Size:
1 pack
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30178672 1 Unit 1 pack
30178671 4 Units Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30178672 Supplier Sartorius Supplier No. SBLRA025EL1
  • ONLINE EXCLUSIVE PRICE
    154.00 €
    145.00 € / Pack of 1
    Save 9.00 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protein A affinity chromatography unit offering a fast, flexible and future-proof solution for antibody purification.

  • High flow rates and reduced residence times take you from equilibration to pure antibody in less than two minutes.
  • Maximum dynamic binding capacity supports the preparation of your antibody sample to the highest possible yield and purity.
  • Ready-for-use units avoid the need for column packing and buffer degassing, while enabling convenient in-line prefiltration.
  • Enhanced pressure resistance in a platform that ensures reliable purification with or without a liquid chromatography system.
  • Future-proof by design, ensuring ease-of-transfer for your most promising targets to clinical and commercial production.
TRUSTED_SUSTAINABILITY

Specifications

Flow Rate 2.5 to 25 mL/min.
Column Type Affinity
Stationary Phase Agarose or Protein A
Diameter (Metric) Inner Column 25 mm
Volume (Metric) 0.5 mL
Quantity 1 Unit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.