Learn More
Sartorius™ Sartopore™2 Capsule 0.2μm Liquid Filters
Ensures highly economic and safe filtration performance. Sartorius™ Sartopore™2 Capsule 0.2μm Liquid Filters provide high-performance polyethersulfone (PES) liquid filtration from single-layer high flow solutions to specific Mycoplasma retentive filters. Available in a broad variety of different pore size combinations.
Brand: Sartorius™ 5441307H4--SS--B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
High-performance polyethersulfone (PES) liquid filters ensure highly economic and safe filtration performance. From single-layer high flow solutions to specific Mycoplasma retentive filters, Sartopore™2 perfectly adapts to your specific process needs. Available in a broad variety of different pore size combinationsSpecifications
Autoclavable | |
PES, Asymmetric | |
Sterile | |
For filtration of biologicals, pharmaceuticals, cell culture media and components and buffers | |
19.05mm Tri-Clamp 25mm | |
5Pack | |
63 mm |
ISO 9001, USP Class VI | |
0.45/0.2 μm | |
0.015 m2 | |
Polypropylene | |
19.05mm Tri-Clamp 25mm | |
Filter Capsule |