Learn More
Sartorius Analytical Draft Shield for Quintix™ and Practum™ Toploader Balances
Shop All Sartorius Products
Click to view available options
:
Draft Ring for Secura Analytical Balances; SQP
Description
Applicable for Quintix or Practum balances with a readability of 10mg and a weighing range of up to 2100g or Quintix/Practum balances with a readability of 0.1g or 1g.
Specifications
Specifications
| For Use With (Equipment) | Quintix or Practum Balance |
| Type | Draft Ring |