Learn More
Sartorius Arium™ Comfort Pre-Filter
Product Code. 15744595 Shop All Sartorius Products
Change view
Click to view available options
For Use With (Equipment):
Arium™ Comfort I and II
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15744595 Arium™ Comfort I and II Each
1 options
Product Code. 15744595 Supplier Sartorius Supplier No. H2OCPFCO1
Equipment and Instrument End of Year Sale
Use promo code 28000
Se alle kampagner
 Restricted Item
In Stock
Læg i indkøbskurv
Edge
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
Arium™ Comfort Pre-Filter for Arium™ comfort water systems.
  • Pre-filter consisting of high efficient activated carbon combined with a process optimizing catalyst
  • Removes reliable oxidizing agents, heavy metals and particular impurities
  • For intended use with: Arium™ Comfort I Arium™ Comfort II
  • Qty. 1 Unit/Pack
TRUSTED_SUSTAINABILITY
Tekniske data
For Use With (Equipment) Arium™ Comfort I and II
Type Pre-treatment Cartridge
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.