missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Offered with multiple MWCOs and two membrane options:
- Cellulose triacetate (CTA) features high hydrophilicity and very low non-specific binding.
- Cast without a support that could bind filtered solutes, these membranes are ideal where high target molecule recovery from the filtrate is important.
- Polyethersulfone (PES) membranes exhibit low fouling characteristics,
- Have a uniform surface, devoid of hydrophobic or hydrophillic interactions, offering excellent protein recoveries for most solutions.
Typical applications for Vivaspin™ Filtrate include:
- Sample preparation for free drug
- Metabolite or hormone analyses
- Sample deproteinization
- Separation of proteins from low molecular weight substances in biological samples
- Sample capacity of 0.5 to 2.5 mL
- May be used in fixed angle or swing bucket rotors, accepting standard conical or flat bottom 15 mL centrifuge tubes
- Typically filter 90% of the initial sample volume in 30 minutes
- Avoid premature membrane blockage by particle loaded samples
- Retrieve the ultrafiltrate immediately after centrifugation without any disassembly
Specifications
Specifications
| Disposable | Single-use |
| Filter Type | Ultrafiltration |
| Material (Membrane) | PES |
| MWCO | 300 kDa |
| Sterility | Non-sterile |
| For Use With (Application) | Small volume centrifugal ultrafiltration of biological samples |
| Max. Force | 2000xG (Fixed Angle Rotor); 2500xG (Swinging Bucket Rotor) |
| Quantity | 12 Pk. |
| Type | Centrifugal Ultrafilter |
| Volume (Metric) | 0.5 to 2.5 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
