Learn More
Sartorius Cellulose Nitrate Membrane Filters
Combines uniform pore structure with high flow and adsorption rates. Sartorius Cellulose Nitrate Membrane Filters are excellently suited for diagnostic kits. Non-sterile, hydrophilic filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
100.00 € - 525.00 €
Specifications
Autoclavable | Autoclavable (wet) |
---|---|
Color | White |
Material | Cellulose Nitrate |
Hydrophobicity | Hydrophilic |
Thickness (Metric) | 130 μm |
Alert:
Cellulose nitrate filters are highly flammable. Their flashpoint is approx. 200°C. Always keep these filters separated by the discs of interleaving paper that are supplied with the filters.
Product Code | Brand | Diameter (Metric) | Pore Size | Flow Rate | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Pore Size | Flow Rate | Price | Quantity & Availability | |||
10303591
|
Sartorius
11301--13------N |
13 mm | 8 μm | 750mL/min. |
115.00 €
Pack of 100 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10272221
|
Sartorius
11301--47------N |
47 mm | 8 μm | 750mL/min. |
204.00 €
Pack of 100 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10068070
|
Sartorius
11303-100------G |
100 mm | 1.2 μm | 200mL/min. |
233.00 €
Pack of 25 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10675051
|
Sartorius
11301--50------N |
50 mm | 8 μm | 750mL/min. |
209.00 €
Pack of 100 |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10383441
|
Sartorius
11303--25------N |
25 mm | 1.2 μm | 200mL/min. |
145.00 €
Pack of 100 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10634751
|
Sartorius
11304--47------N |
47 mm | 0.8 μm | 200mL/min. |
194.00 €
Pack of 100 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10231651
|
Sartorius
11305--47------N |
47 mm | 0.65 μm | 130mL/min. |
194.00 €
Pack of 100 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10262131
|
Sartorius
11304--25------N |
25 mm | 0.8 μm | 200mL/min. |
145.00 €
Pack of 100 |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10494261
|
Sartorius
11305--25------N |
25 mm | 0.65 μm | 130mL/min. |
145.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10333491
|
Sartorius
11306-100------G |
100 mm | 0.45 μm | 70mL/min. |
179.00 €
Pack of 25 |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10193891
|
Sartorius
11306--90------N |
90 mm | 0.45 μm | 70mL/min. |
525.00 €
Pack of 100 |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10567711
|
Sartorius
11306--25------N |
25 mm | 0.45 μm | 70mL/min. |
110.00 €
Pack of 100 |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10425401
|
Sartorius
11306--50------N |
50 mm | 0.45 μm | 70mL/min. |
130.00 €
Pack of 100 |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12972661
|
Sartorius
11306--47------N |
47 mm | 0.45 μm | 70mL/min. |
130.00 €
Pack of 100 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10008060
|
Sartorius
11306--90------G |
90 mm | 0.45 μm | 70mL/min. |
165.00 €
Pack of 25 |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10199130
|
Sartorius
11342--25------N |
25 mm | 5 μm | 570mL/min. |
182.00 €
Pack of 100 |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10508871
|
Sartorius
11304--50------N |
50 mm | 0.8 μm | 200mL/min. |
211.00 €
Pack of 100 |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10404651
|
Sartorius
11302--25------N |
25 mm | 3 μm | 430mL/min. |
145.00 €
Pack of 100 |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10211751
|
Sartorius
11302--47------N |
47 mm | 3 μm | 430mL/min. |
198.00 €
Pack of 100 |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10018070
|
Sartorius
11306--47------N |
47 mm | 0.45 μm | 70mL/min. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Combines uniform pore structure with high flow and adsorption rates. Sartorius Cellulose Nitrate Membrane Filters are excellently suited for diagnostic kits. Non-sterile, hydrophilic filters in a variety of diameters and pore sizes are bulk-packed to save money and space.
- Cellulose nitrate filters combine high flow rates with high non-specific adsorption
- Defined particle retention and outstanding, reliable capture rates for microorganisms
- Very advantageous for diagnostic kits and compatible with aqueous solutions, hydrocarbons and several other organic solvents
- Filters have a uniform pore structure and are available in range of diameters and pore sizes
- Pore sizes: 0.45μm, 0.65μm, 0.8μm, 1.2μm, 3.0μm, 5μm and 8μm
- The larger pore sizes (8 μm, 5 μm, 3 μm) are useful for cell retention and prefiltering
- The 0.45 pore size is ideal for particle collection and filtration
- Diameters: 13, 25, 37, 47, 50, 90 and 100mm (0.5 to 3.9 in.)
- Ungridded filters can be sterilized via autoclave (wet) to 121°C, ethylene oxide or γ-irradiation to 25 kGy
- Box label specifies the type, quantity, diameter, pore size and lot number
Specifications
Autoclavable (wet) | |
Cellulose Nitrate | |
130 μm |
White | |
Hydrophilic |