Learn More
Sartorius Interface Module for Cubis Balances
Interface Module for use with Cubis balances.
Brand: Sartorius YDO01MS-P
Code : NEW
Additional Details : Weight : 4.00000kg
Description
- Option R (YDO01MS-R): RS-232C data interface, 25-pin, for connection of Cubis accessory equipment
- Option B (YDO01MS-B): Bluetooth™ interface module for wireless connection of the YDP10BT data printer
- Option P (YDO01MS-P): RS-232C data interface, 9-pin, including PS/2 port for connecting a PC or a keyboard
Specifications
Cubis Balance | |
Option P: RS-232 9 Pin, PS/2 | |
Connecting a PC or a keyboard | |
Data Interface |