Learn More
Sartorius Support Arm, for Cubis™ High-Capacity Balances
GREENER_CHOICE
Product Code. 15457160 Shop All Sartorius Products
Click to view available options
:
Support Arm for Cubis High Capacity Precision Weighing Modules
Product Code. 15457160
Brand: Sartorius YDH02MS
Sturdy support arm for Cubis high-capacity balances.
For precision weighing modules with 100mg/1g readability and weighing capacity ≥20kg; for raising the MSE, MSU, and MSA display and control units. With the display raised on a support arm, you can set up your balance so that even working with heavy loads is ergonomic and minimally stressful.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cubis High Capacity Balance
Type Support Arm