missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Density Kits, for Secura™, Quintix™ and Practum™ Analytical Balances
GREENER_CHOICE

Product Code. 15199976 Shop All Sartorius Products
Click to view available options
:
Density Kit; for Secura, Quintix, Practum analytical balances
This item is not returnable. View return policy

Product Code. 15199976

Brand: Sartorius YDK03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Secura™, Quintix™ and Practum Analytical Balances.

For use with Secura™, Quintix™ and Practum Analytical Balances.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Secura, Quintix, Practum Analytical Balance
Type Density Determination Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.