Learn More
Sartorius Minisart™ HY Syringe Filters
Designed for fast and reliable sterile venting and gas filtration. Sartorius Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0μm pore sizes. Sterile option available.
Brand: Sartorius 16596--------HYK
170.05 EUR valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Code : 53
Additional Details : Weight : 0.36900kg
Description
Designed for fast and reliable sterile venting and gas filtration. Sartorius Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0μm pore sizes. Sterile option available.
Fast and reliable sterile venting and filtration- Clean, safe filters that are almost free of leachables and extractables
- Reliably removes particles and microorganisms with no leakage
- Has minimal effect on the ingredients of the filtered solution
- Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
- Wide variety of lab uses:
- Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
- Sterile venting of small fermenters or bioreactors and of containers for culture media
- Also used for aqueous and oily ear- or eye-solutions and other drugs
- Complete units are ready to use
- Single use/disposable filter units
- Female luer lock inlet connectors with male leur lock outlet connector
- Proven quality and 100% tested during manufacture
- Sterile (via ethylene oxide) and non-sterile units available
Specifications
Clear | |
PTFE | |
Sterile | |
0.25 mL | |
Hydrophobic | |
Male Luer Lock | |
50/Pk. | |
Syringe Filter |
Single-use | |
0.2 μm | |
5.16 cm2 | |
Medical acrylic (MBS) | |
Female Luer Lock | |
4.5 bar | |
Cryolite | |
26 mm |