missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Minisart™ HY Syringe Filters
Designed for fast and reliable sterile venting and gas filtration. Sartorius Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0μm pore sizes. Sterile option available.
179.00 € - 1070.00 €
Specifications
Color | Clear |
---|---|
Disposable | Single-use |
Material | PTFE |
Filtration Area | 5.16 cm2 |
Housing Material | Medical acrylic (MBS) |
Product Code | Brand | Pore Size | Sterility | Hold up Volume (After Air Purge) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Sterility | Hold up Volume (After Air Purge) | Price | Quantity & Availability | |||
10695211
|
Sartorius
16596--------HYK |
0.2 μm | Sterile | 0.25 mL |
179.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
10304941
|
Sartorius
16596--------HYQ |
0.2 μm | Non-sterile | 0.25 mL |
1070.00 €
Pack of 500 |
In Stock
Log in to see stock. |
|||
Description
Designed for fast and reliable sterile venting and gas filtration. Sartorius Minisart™ HY Syringe Filters are single-use, complete filtration units with a naturally hydrophobic PTFE membrane and medical acrylic housing. Choose from 0.2 and 1.0μm pore sizes. Sterile option available.
Fast and reliable sterile venting and filtration- Clean, safe filters that are almost free of leachables and extractables
- Reliably removes particles and microorganisms with no leakage
- Has minimal effect on the ingredients of the filtered solution
- Includes a naturally hydrophobic, 26mm PTFE membrane with medical acrylic (MBS) housing
- Wide variety of lab uses:
- Reliable filtration of particulates and cells from gasses, additives, buffers and reagents
- Sterile venting of small fermenters or bioreactors and of containers for culture media
- Also used for aqueous and oily ear- or eye-solutions and other drugs
- Complete units are ready to use
- Single use/disposable filter units
- Proven quality and 100% tested during manufacture
- Sterile (via ethylene oxide) and non-sterile units available
Specifications
Clear | |
PTFE | |
Medical acrylic (MBS) | |
4.5 bar | |
Syringe Filter |
Single-use | |
5.16 cm2 | |
Hydrophobic | |
Cryolite | |
26 mm |