missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Nutrient Pad Sets (NPS) for Microsart™ e.motion Dispenser

Product Code. 11790656 Shop All Sartorius Products
Change view
Click to view available options
Media Type:
Azide
Cetrimide
Endo
Orange Serum
Schaufus Pottinger
Schaufus Pottinger (m-Green Yeast and Mold)
Standard TTC
TGE
Tergitol TTC
Wort
Diameter (Metric) Outside:
4.7 cm
5 cm
Pore Size:
0.45μm
0.65μm
0.8μm
Unit Size:
Pack of 100
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Media Type Diameter (Metric) Outside Pore Size Unit Size
11790656 TGE 4.7 cm 0.45μm Pack of 100
11740646 Wort 5 cm 0.65μm Pack of 100
11873272 Endo 5 cm 0.45μm Pack of 100
11823272 Azide 4.7 cm 0.45μm Pack of 100
11803282 Standard TTC 5 cm 0.45μm Pack of 100
11813282 Tergitol TTC 5 cm 0.45μm Pack of 100
11883272 Orange Serum 4.7 cm 0.45μm Pack of 100
15922490 Tergitol TTC 4.7 cm 0.45μm Pack of 100
15942490 Orange Serum 5 cm 0.45μm Pack of 100
11843272 Cetrimide 4.7 cm 0.45μm Pack of 100
11853272 Cetrimide 5 cm 0.45μm Pack of 100
15922500 Schaufus Pottinger (m-Green Yeast and Mold) 5 cm 0.8μm Pack of 100
11863272 Endo 4.7 cm 0.45μm Pack of 100
11893272 Standard TTC 4.7 cm 0.45μm Pack of 100
11720646 Wort 4.7 cm 0.65μm Pack of 100
11710666 TGE 5 cm 0.45μm Pack of 100
15912500 Schaufus Pottinger 5 cm 0.8μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 11790656 Supplier Sartorius Supplier No. 1407647RDN
  • ONLINE EXCLUSIVE PRICE
    265.00 €
    224.55 € / Pack of 100
    Save 40.45 €
    15% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sterile, dehydrated culture media which is individually inserted in a petri dish and sterilized

Sartorius Nutrient Pad Sets are sterile, dehydrated culture media designed for use with membrane filter methods. Each Nutrient Pad Set includes the appropriate membrane filters (in 47 or 50mm diameters), which are also sterile and individually packaged. Just moisten the Nutrient Pads with sterile and demineralized (or distilled) water and they are immediately ready to use. They comply with international regulations and recommendations including, but not limited to: international pharmacopoeias, DIN and ISO standards, the American Standards for Water and Foods, mineral water regulations, brewery guidelines, such as MEBAC or EBC, and recommendations of the food industry, such as LMBG, NCA and ICUMSA.

  • Trouble-free storage up to 24 months
  • Economical -- time and cost savings
  • Easy handling
  • Validation guide available
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance DIN and ISO standards
Color Green
Form Dehydrated
Manufacturing Origin Germany
Media Type TGE
Product Type Nutrient Pad
Sterility Sterile
Test Type Microbial Detection
For Use With (Application) Total colony count
Grid Color Dark Green
Packaging Type Individually Packaged
Pore Size 0.45μm
Diameter (English) Outside 1.85 in.
Diameter (Metric) Outside 4.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.