Learn More
Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
Product Code. 11790956 Shop All Sartorius Products
Change view
Click to view available options
For Use With:
Vivacell 70 pressure fuge
Vivacell™ 100
Vivacell™ 250
Vivacell™ 70
Vivaspin™ 20
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11790956 Vivaspin™ 20 Each
11705036 Vivacell™ 250 Each
10537441 Vivacell™ 70 Each
11735036 Vivacell 70 pressure fuge Each
10609731 Vivacell™ 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11790956 Supplier Sartorius Supplier No. VCA200
  • ONLINE EXCLUSIVE PRICE
    105.00 €
    97.59 € / Pack of 1
    Save 7.41 €
    7% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For various Vivacell models
The Vivaspin™ 20 is an ultrafiltration unit with unique twin membrane for sample concentration with high process flexibility: centrifugation or centrifugation of pressurized units, as well as pressure filtration in stationary benchtop mode.
TRUSTED_SUSTAINABILITY
Specifications
For Use With Vivaspin™ 20
Type Pressure Head