Learn More
Sartorius Pressure Heads for Vivaspin™ 20 and Vivacell™ 70, 100, 250
For various Vivacell models
127.00 € - 340.00 €
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
11790956
|
Sartorius
VCA200 |
Vivaspin™ 20 |
127.00 €
Each |
Estimated Shipment: 21-06-2024
Log in to see stock. |
|||||
10609731
|
Sartorius
VCA800 |
Vivacell™ 100 |
340.00 €
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||||
11705036
|
Sartorius
VCA015 |
Vivacell™ 250 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11735036
|
Sartorius
VCA701 |
Vivacell™ 70 pressure fuge |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10537441
|
Sartorius
VCA700 |
Vivacell™ 70 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Vivaspin™ 20 is an ultrafiltration unit with unique twin membrane for sample concentration with high process flexibility: centrifugation or centrifugation of pressurized units, as well as pressure filtration in stationary benchtop mode.Vivacell™ 100 is a disposable ultrafiltration unit for the concentration of samples up to 98 mL that can be used either in a centrifuge or for pressure filtration. Vivacell™ 100 thus combines the ease of use of centrifugal devices with the flexibility and control provided by pressurized ultrafiltration cells.Specifications
Pressure Head |