Learn More
Descrizione
Vivapure miniprep and spin columns are pre-packed centrifugal devices for purification of monoclonal and polyclonal antibodies from serum, ascites and cell culture supernatants. The rapid 1-2-3 bind-wash-elute protocol especially lends itself to screening applications, where many samples are processed in parallel. Vivapure miniprep & spin columns can conveniently be used in a centrifuge with swing-out rotor for standard 2.2 ml tubes. Binding capacity is about 1 mg IgG per unit. The low hold-up volume ensures high solute recovery with minimal loss by non-specific adsorption. Vivapure miniprep and spin columns can be re-used three times and are available as complete kits including buffers or as bulk packs.They are an attractive alternative to lengthy chromatographic methods such as FPLC. For larger volumes, the Vivapure Maxiprep kit is also availablity.
- Spin column based kit
- Devices are ready to use
- Rapid purification with high yields
- Parallel processing
- Cost effective through re-use option
- Starter kits available
Specifica
Specifica
| Matrix | Protein A Resin |
| Quantity | Each |
| For Use With (Application) | Purification of Monoclonal and Polyclonal Antibodies from Serum, ascites and cell culture supernatants |
| Type | Miniprep Spin Column |
| Sufficient For | 4 Maxi or 48 miniprep Protein A and G spin column |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.