Learn More
Sartorius Vivaspin™ 500 Centrifugal Concentrators
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
Brand: Sartorius VS0161
Code : 50
Additional Details : Weight : 0.10000kg
Description
Provides a simple, one-step concentration procedure. Sartorius Vivaspin™ 500 Centrifugal Concentrators are disposable ultrafiltration devices for the concentration of biological samples with starting volumes up to 500μL. Available with PES membranes in a wide range of molecular weight cut-offs.
- Patented vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides high speed concentrations
- Concentrate recoveries of > 90% can be reached in less than 30 minutes
- The dead stop pocket prevents concentration to dryness for safe sample concentration
- Universal rotor compatibility: can be used in fixed angle rotors accepting 2.2mL centrifuge tubes
- High-flux PES membranes for ultra-fast concentration
- Convenient concentrate recovery by pipetting
- Clear housing with volume graduation for easy reading/monitoring
Specifications
Single-use | |
1000 kDa | |
Polycarbonate | |
Centrifugal Concentrator | |
50 mm |
PES | |
64.5m m2 | |
25/Pk. | |
500 μL |