missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Vivaspin™ 6 Centrifugal Concentrators
BIOPHARMA

Product Code. 11741826 Shop All Sartorius Products
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
1000 kDa
3 kDa
30 kDa
300 kDa
5 kDa
5, 10, 30, 50, and 100 kDa
50 kDa
Quantity:
100/Pk.
25/Pk.
Unit Size:
Pack of 100
Pack of 25
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Quantity Unit Size
11741826 300 kDa 25/Pk. Pack of 25
16656752 10 kDa 25/Pk. Pack of 25
10421243 10 kDa 100/Pk. Pack of 100
16676752 10 kDa 100/Pk. Pack of 100
10767461 5 kDa 25/Pk. Pack of 25
10293471 5 kDa 100/Pk. Pack of 100
10029460 30 kDa 25/Pk. Pack of 25
10384981 30 kDa 100/Pk. Pack of 100
10395021 50 kDa 25/Pk. Pack of 25
10588191 50 kDa 100/Pk. Pack of 100
10120901 100 kDa 100/Pk. Pack of 100
10009640 100 kDa 25/Pk. Pack of 25
11740746 300 kDa 100/Pk. Pack of 100
10777461 1000 kDa 25/Pk. Pack of 25
10110951 1000 kDa 100/Pk. Pack of 100
10059520 - 25/Pk. Pack of 25
10449512 - 100/Pk. Pack of 100
11770746 3 kDa 100/Pk. Pack of 100
10150203 3 kDa 25/Pk. Pack of 25
10738421 5, 10, 30, 50, and 100 kDa 25/Pk. Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
This item is not returnable. View return policy
Product Code. 11741826 Supplier Sartorius Supplier No. VS0651
  • ONLINE EXCLUSIVE PRICE
    258.00 €
    240.00 € / Pack of 25
    Save 18.00 €
    6% Off
View all promotions
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Achieve fast sample concentration and high recovery. Sartorius Vivaspin™ 6 Centrifugal Concentrators are the only device of its size that can process 6mL samples. They feature twin vertical membranes and built-in dead stop pockets. Available with PES membranes in a wide range of molecular weight cut-offs.

Developed to offer increased volume flexibility and performance
  • Compact tubes can process up to an impressive 6mL initial volume
  • Optimally suited for the concentration of antibody, protein, nanoparticle and virus samples
  • Universal rotor compatibility:
    • Use in either swing bucket or fixed angle rotors accepting standard 15mL conical bottom test tubes
Fast, safe and easy concentration
  • Features premium, high-throughput PES membranes
  • Twin vertical membrane design lead to unparalleled filtration speed and over 100-fold concentration
  • The dead stop pocket prevents concentration to dryness for safe sample concentration
  • Nearly complete concentrate recovery of over 90%
  • Remaining volume is easy to see with the printed graduations and clear housing
  • Modified dead stop pocket further simplifies direct pipette recovery of the final concentrate
TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Filter Type Ultrafiltration
Material (Membrane) PES
MWCO 300 kDa
Sterility Non-sterile
Filtration Area 2.58 cm2
Housing Material Polycarbonate
Quantity 25/Pk.
Type Centrifugal Concentrators
Volume (Metric) 6 mL
Length (Metric) 122 mm
For Use With (Equipment) Centrifuge
Sample Volume 6 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.